Glutaric anhydride ≥95%

Synonyms: Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione

G3806-5G G3806-100G G3806-500G
220179-165EA 51.65 USD
220179-165 220179-163 220179-164
Glutaric anhydride ≥95%
Glutaric anhydride
Formula: C₅H₆O₃
MW: 114.1 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 52…55 °C
Density: 1.411 g/cm³ (20 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006679
CAS Number: 108-55-4
EINECS: 203-593-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR