D-(+)-Galactose

Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal

G0625-100G G0625-5KG G0625-500G G0625-1KG
220178-502EA 56.25 USD
220178-502 220178-505 220178-504 220178-503
D-(+)-Galactose
D-(+)-Galactose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 527 °C (1013 hPa)
Melting Pt: 163…169 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR