Fuchsin (acid)

Synonyms: Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)

F8129-50G F8129-100G F8129-25G
220177-783EA 203 USD
220177-783 220177-781 220177-782
Fuchsin (acid)
Fuchsin (acid)
Formula: C₂₀H₁₇N₃Na₂O₉S₃
MW: 585.55 g/mol
Melting Pt: ? 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00013286
CAS Number: 3244-88-0
EINECS: 221-816-5
Merck Index: 12,00109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4018bf04429ebc2757c43d7c7a9de213ceef8186&vid=3dfb10465fa2d34a2f4b414e7f6e92b1f7ea2f15&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR