5-Fluorocytosine

Synonyms: 5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one

F7129-1G F7129-5G
220177-673EA 330.6 USD
220177-673 220177-674
5-Fluorocytosine
5-Fluorocytosine
Formula: C₄H₄FN₃O
MW: 129.09 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 165 °C
MDL Number: MFCD00006035
CAS Number: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR