3-Fluorobenzoic acid ≥97%

Warning

F6605-25G F6605-5G
220177-612EA 52.34 USD
220177-612 220177-613
3-Fluorobenzoic acid ≥97%
3-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS Number: 455-38-9
EINECS: 207-248-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR