Ferrocene

F408-500G F408-100G F408-5G
220176-381EA 161.23 USD
220176-381 220176-380 220176-382
Ferrocene
Ferrocene
Formula: C₁₀H₁₀Fe
MW: 186.04 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 172…175 °C
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00001427
CAS Number: 102-54-5
EINECS: 203-039-3
UN: 1325
ADR: 4.1,II
Merck Index: 13,04070

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR