4-Fluoroaniline ≥99%

Warning

F3800-100G F3800-25G F3800-500G
220177-363EA 48.46 USD
220177-363 220177-364 220177-365
4-Fluoroaniline ≥99%
4-Fluoroaniline
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.16 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007829
CAS Number: 371-40-4
EINECS: 206-735-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,04196

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR