2-Fluoroaniline ≥99%

Warning

F3401-100G-A F3401-25G-A F3401-500G-A
220177-235EA 140.77 USD
220177-235 220177-236 220177-237
2-Fluoroaniline ≥99%
2-Fluoroaniline
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007642
CAS Number: 348-54-9
EINECS: 206-478-9
UN: 2941
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99606041da3b710b3ce46eb7ee3213bc4a2f5090&vid=99606041da3b710b3ce46eb7ee3213bc4a2f5090&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a536f229937fe777bec28569bff335e83a3ac6ac&vid=a536f229937fe777bec28569bff335e83a3ac6ac&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10c6b64e20ee238295e5e32a1695b5d45fd34daa&vid=10c6b64e20ee238295e5e32a1695b5d45fd34daa&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR