4'-Fluoroacetophenone ≥99%

Warning

Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone

F3207-25G F3207-100G
220177-212EA 56.32 USD
220177-212 220177-211
4'-Fluoroacetophenone ≥99%
4'-Fluoroacetophenone
Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 196 °C (1013 hPa)
Density: 1.138 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000354
CAS Number: 403-42-9
EINECS: 206-960-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR