Iron(III) acetylacetonate

Warning

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

F300-25G F300-100G F300-500G
220176-476EA 60.62 USD
220176-476 220176-475 220176-477
Iron(III) acetylacetonate
Iron(III) acetylacetonate
Formula: C₁₅H₂₁FeO₆
MW: 353.18 g/mol
Boiling Pt: 100 °C (9.7513 mmHg)
Melting Pt: 180…183 °C
Density: 5.24 g/cm³ (20 °C)
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR