2-Fluorotoluene ≥99%

Synonyms: 1-Fluoro-2-methylbenzene, o-Fluorotoluene

F15323-100G F15323-500G
220176-953EA 60.31 USD
220176-953 220176-954
2-Fluorotoluene ≥99%
2-Fluorotoluene
Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 113 °C (1013 hPa)
Melting Pt: –62 °C
Density: 1.005 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00000322
CAS Number: 95-52-3
EINECS: 202-428-5
UN: 2388
ADR: 3,II
Merck Index: 13,04207

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR