4-Fluorothiophenol ≥98%

Danger

Synonyms: 4-Fluorobenzene-1-thiol, 1-Fluoro-4-mercaptobenzene, 4-Fluorobenzenethiol, p-Fluorothiophenol

F15315-10G F15315-25G
220176-951EA 80.34 USD
220176-951 220176-952
4-Fluorothiophenol ≥98%
4-Fluorothiophenol
Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 164 °C (1013 hPa)
Density: 1.1996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004846
CAS Number: 371-42-6
EINECS: 206-737-6
UN: 3071
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR