2-Fluoropyridine

Danger

F15250-10G F15250-100G
220176-949EA 38.45 USD
220176-949 220176-948
2-Fluoropyridine
2-Fluoropyridine
Formula: C₅H₄FN
MW: 97.09 g/mol
Boiling Pt: 126 °C (1013 hPa)
Density: 1.129 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006224
CAS Number: 372-48-5
EINECS: 206-757-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR