(4-Fluorophenyl)acetic acid ≥98%

Synonyms: 2-(4-Fluorophenyl)acetic acid

F13304-25G F13304-100G
220175-837EA 59.12 USD
220175-837 220175-836
(4-Fluorophenyl)acetic acid ≥98%
(4-Fluorophenyl)acetic acid
Formula: (C₆H₄F)CH₂COOH
MW: 154.14 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 84…85 °C
MDL Number: MFCD00004343
CAS Number: 405-50-5
EINECS: 206-972-4
Merck Index: 13,04204

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR