4-Fluorophenol ≥99%

Warning

Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene

F13207-100G F13207-25G
220175-833EA 106.83 USD
220175-833 220175-834
4-Fluorophenol ≥99%
4-Fluorophenol
Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 44…47 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002316
CAS Number: 371-41-5
EINECS: 206-736-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR