3-Fluorophenol ≥98%

Warning

F13002-10G F13002-100G F13002-50G
220176-091EA 83.25 USD
220176-091 220176-934 220175-823
3-Fluorophenol ≥98%
3-Fluorophenol
Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: 13.7 °C
Density: 1.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002254
CAS Number: 372-20-3
EINECS: 206-748-6
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR