2-Fluorophenol ≥98%

Danger

Synonyms: o-Fluorophenol

F12804-100G F12804-10G
220176-930EA 188.46 USD
220176-930 220176-931
2-Fluorophenol ≥98%
2-Fluorophenol
Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002155
CAS Number: 367-12-4
EINECS: 206-681-2
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR