5-Fluoro-2-nitrotoluene ≥97%

Warning

Synonyms: 1-Fluoro-3-methyl-4-nitrobenzene

F12405-50G F12405-10G
220176-924EA 73.72 USD
220176-924 220176-923
5-Fluoro-2-nitrotoluene ≥97%
5-Fluoro-2-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007283
CAS Number: 446-33-3
EINECS: 207-164-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR