1-Fluoro-4-nitrobenzene ≥99%

Synonyms: 4-Fluoronitrobenzene, 1-Fluoro-4-nitrobenzene

F11204-100G F11204-500G F11204-5G
220176-895EA 64.62 USD
220176-895 220176-896 220176-897
1-Fluoro-4-nitrobenzene ≥99%
1-Fluoro-4-nitrobenzene
Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 21 °C
Density: 1.326 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007282
CAS Number: 350-46-9
EINECS: 206-502-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR