1-Fluoro-2-nitrobenzene ≥99%

Warning

Synonyms: o-Fluoronitrobenzene, 2-Fluoronitrobenzene

F10801-250G F10801-50G
220176-891EA 141.08 USD
220176-891 220176-892
1-Fluoro-2-nitrobenzene ≥99%
1-Fluoro-2-nitrobenzene
Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007048
CAS Number: 1493-27-2
EINECS: 216-088-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR