Ethyl acrylate ≥99%

Synonyms: Acrylic acid-ethyl ester

E9706-100ML E9706-1L E9706-2L
220124-858EA 61.06 USD
220124-858 220124-859 220124-860
Ethyl acrylate ≥99%
Ethyl acrylate
Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00009188
CAS Number: 140-88-5
EINECS: 205-438-8
UN: 1917
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR