Ethanolamine ≥98%

Danger

Synonyms: 2-Aminoethanol, Monoethanolamine

E9508-2.5L E9508-1L E9508-500ML E9508-100ML
220122-689EA 124.06 USD
220122-689 220122-688 220122-690 220122-686
Ethanolamine ≥98%
Ethanolamine
Formula: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: 10.5 °C
Density: 1.0164 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8,III
Merck Index: 13,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR