EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)

Warning

Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl

E7750-1G E7750-10G E7750-1KG E7750-100G E7750-100MG E7750-5G E7750-5KG E7750-25G
220124-666EA 34.38 USD
220124-666 220124-665 220124-667 220124-663 220124-664 220124-669 220124-670 220124-668
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Formula: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Melting Pt: 112 - 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR