Eugenol ≥99%

Warning

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

E51791-100G E51791-500G E51791-5G
220122-535EA 50.72 USD
220122-535 220122-536 220122-537
Eugenol ≥99%
Eugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: ∼–10 °C
Density: 1.0652 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR