1-Ethynylcyclohexanol ≥99%

Synonyms: 1-Ethynyl-1-cyclohexanol

E51406-20L E51406-100ML E51406-5L
220124-292EA 5276.92 USD
220124-292 220124-291 220124-293
1-Ethynylcyclohexanol ≥99%
1-Ethynylcyclohexanol
Formula: C₈H₁₂O
MW: 124.18 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 0.973 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003858
CAS Number: 78-27-3
EINECS: 201-100-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR