(Ethyl)triphenylphosphonium bromide

Warning

Synonyms: Ethyltriphenylphosphonium bromide

E50604-25G E50604-100G
220124-276EA 46.31 USD
220124-276 220124-275
(Ethyl)triphenylphosphonium bromide
(Ethyl)triphenylphosphonium bromide
Formula: C₂₀H₂₀BrP
MW: 371.26 g/mol
Melting Pt: 205 - 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00011838
CAS Number: 1530-32-1
EINECS: 216-223-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR