Ethyl-4,4,4-trifluoroacetoacetate ≥99%

Synonyms: Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate

E50205-25G E50205-5G E50205-100G
220124-268EA 55.57 USD
220124-268 220124-269 220124-267
Ethyl-4,4,4-trifluoroacetoacetate ≥99%
Ethyl-4,4,4-trifluoroacetoacetate
Formula: C₆H₇F₃O₃
MW: 184.11 g/mol
Boiling Pt: 131 °C (1013 hPa)
Melting Pt: -39 °C
Density: 1.2586 g/cm³ (15.5 °C)
MDL Number: MFCD00000424
CAS Number: 372-31-6
EINECS: 206-750-7
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR