o-Ethyltoluene ≥99%

Synonyms: 1-Ethyl-2-methylbenzene, 2-Ethyltoluene

E49401-5G E49401-25G
220124-255EA 124.92 USD
220124-255 220124-254
o-Ethyltoluene ≥99%
o-Ethyltoluene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 164 °C (1013 hPa)
Melting Pt: -17 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009257
CAS Number: 611-14-3
EINECS: 210-255-1
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR