Ethyl pyruvate ≥98%

Synonyms: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester

E47808-25G E47808-100G
220124-230EA 40.06 USD
220124-230 220124-229
Ethyl pyruvate ≥98%
Ethyl pyruvate
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 144…147 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009123
CAS Number: 617-35-6
EINECS: 210-511-2
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR