Ethyl propiolate ≥99%

Synonyms: Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester

E46607-100G E46607-5G E46607-25G
220124-210EA 370.46 USD
220124-210 220124-212 220124-211
Ethyl propiolate ≥99%
Ethyl propiolate
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: 9 °C
Density: 0.9788 g/cm³ (4 °C)
MDL Number: MFCD00009184
CAS Number: 623-47-2
EINECS: 210-795-8
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR