2-Ethylphenol ≥99%

Synonyms: 1-Ethyl-2-hydroxybenzene

E44000-100G E44000-25G E44000-5G
220124-166EA 212.15 USD
220124-166 220124-167 220124-168
2-Ethylphenol ≥99%
2-Ethylphenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 204.5 °C (1013 hPa)
Melting Pt: –18 °C
Density: 1.0371 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002249
CAS Number: 90-00-6
EINECS: 201-958-4
UN: 3145
ADR: 8,III
Merck Index: 13,03871

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR