Ethyl nicotinate

Synonyms: 3-Pyridinecarboxylic acid ethyl ester, Ethyl 3-pyridinecarboxylate, Pyridine-3-carboxylic acid ethyl ester, 3-Picolinic acid ethyl ester, Nicotinic acid ethyl ester

E40609-100G E40609-500G
220123-077EA 65.69 USD
220123-077 220123-078
Ethyl nicotinate
Ethyl nicotinate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 223 °C (1013 hPa)
Density: 1.115 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006389
CAS Number: 614-18-6
EINECS: 210-370-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR