Ethanethiol ≥97%

Danger

Synonyms: Ethyl mercaptan, 1-Mercaptoethane

E3708-500ML E3708-100ML
220124-116EA 104.78 USD
220124-116 220124-115
Ethanethiol ≥97%
Ethanethiol
Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: -148 °C
Density: 0.846 g/cm³ (20 °C)
Flash Pt: -45 °C
Storage Temperature: Ambient
MDL Number: MFCD00004887
CAS Number: 75-08-1
EINECS: 200-837-3
UN: 2363
ADR: 3,I
Merck Index: 12,03771

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR