2-Ethyl-4-methylimidazole

Synonyms: 2-Ethyl-4-methyl-1H-imidazole

E36652-100G E36652-5G
220124-113EA 80.23 USD
220124-113 220124-114
2-Ethyl-4-methylimidazole
2-Ethyl-4-methylimidazole
Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 45 °C
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00005193
CAS Number: 931-36-2
EINECS: 213-234-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR