Ethyl-2-methylacetoacetate ≥90%

Synonyms: Ethyl 2-methyl-3-oxobutanoate

E35400-25G E35400-100G
220123-430EA 51.58 USD
220123-430 220123-429
Ethyl-2-methylacetoacetate ≥90%
Ethyl-2-methylacetoacetate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.0238 g/cm³ (15.3 °C)
MDL Number: MFCD00009164
CAS Number: 609-14-3
EINECS: 210-179-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR