Ethyl-(S)-(-)-lactate ≥98%

Danger

Synonyms: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate

E34102-500G E34102-100G E34102-5G
220123-416EA 70 USD
220123-416 220123-415 220123-417
Ethyl-(S)-(-)-lactate ≥98%
Ethyl-(S)-(-)-lactate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 154 °C (1013 hPa)
Melting Pt: -25 °C
Density: 1.042 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00004518
CAS Number: 687-47-8
EINECS: 211-694-1
UN: 1192
ADR: 3,III
Merck Index: 12,03850

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR