Ethyl isothiocyanate ≥97%

E33904-50G E33904-10G
220124-097EA 301.54 USD
220124-097 220124-096
Ethyl isothiocyanate ≥97%
Ethyl isothiocyanate
Formula: C₃H₅NS
MW: 87.15 g/mol
Boiling Pt: 130…132 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004820
CAS Number: 542-85-8
EINECS: 208-831-2
UN: 2929
ADR: 6.1,II
Merck Index: 13,03848

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR