Ethyl indole-2-carboxylate

Synonyms: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

E32800-25G E32800-5G
220124-073EA 217.54 USD
220124-073 220124-074
Ethyl indole-2-carboxylate
Ethyl indole-2-carboxylate
Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD00005609
CAS Number: 3770-50-1
EINECS: 223-206-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR