2-Ethylhexane-1,3-diol (mixture of isomers) ≥97%

Synonyms: Ethoexadiol, Octylene glycol

E29125-1KG E29125-500G
220124-021EA 132.46 USD
220124-021 220124-022
2-Ethylhexane-1,3-diol (mixture of isomers) ≥97%
2-Ethylhexane-1,3-diol (mixture of isomers)
Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.9422 g/cm³ (20 °C)
MDL Number: MFCD00004578
CAS Number: 94-96-2
EINECS: 202-377-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR