Ethyl-4-(dimethylamino)benzoate ≥99%

Synonyms: 4-(Dimethylamino)benzoic acid ethyl ester, Ethyl 4-dimethylaminobenzoate

E24905-500G E24905-100G E24905-5G
220123-939EA 443.31 USD
220123-939 220123-938 220123-940
Ethyl-4-(dimethylamino)benzoate ≥99%
Ethyl-4-(dimethylamino)benzoate
Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Boiling Pt: 112 °C (0.02 -rr)
Melting Pt: 63 - 65 °C
Density: 1.0099 g/cm³ (99.7 °C)
MDL Number: MFCD00009115
CAS Number: 10287-53-3
EINECS: 233-634-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR