Ethyl cyanoacetate ≥98%

E18425-250G E18425-1KG E18425-5G
220122-531EA 57.72 USD
220122-531 220122-530 220122-532
Ethyl cyanoacetate ≥98%
Ethyl cyanoacetate
Formula: C₅H₇NO₂
MW: 113.12 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.063 g/cm³ (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00001940
CAS Number: 105-56-6
EINECS: 203-309-0
Merck Index: 13,03821

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR