Ethyl-2-chloroacetoacetate ≥95%

Synonyms: Ethyl 2-chloroacetoacetate

E16902-100G E16902-500G
220122-515EA 56.77 USD
220122-515 220122-516
Ethyl-2-chloroacetoacetate ≥95%
Ethyl-2-chloroacetoacetate
Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: –80 °C
Density: 1.19 g/cm³ (14 °C)
MDL Number: MFCD00009141
CAS Number: 609-15-4
EINECS: 210-180-4

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR