Ethyl butyrate ≥99%

Synonyms: Butanoic acid ethyl ester, Butyric acid ethyl ester

E15701-500ML E15701-25ML E15701-1L
220123-796EA 73.88 USD
220123-796 220123-795 220123-794
Ethyl butyrate ≥99%
Ethyl butyrate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: -92 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009394
CAS Number: 105-54-4
EINECS: 203-306-4
UN: 1180
ADR: 3,III
Merck Index: 13,03810

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR