4-Ethylaniline ≥98%

E12001-500ML E12001-100ML
220123-712EA 327.38 USD
220123-712 220123-711
4-Ethylaniline ≥98%
4-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.96875 g/cm³ (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00007921
CAS Number: 589-16-2
EINECS: 209-637-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR