2-Ethylaniline ≥98%

Warning

E11803-250G E11803-100G
220123-710EA 77.32 USD
220123-710 220123-709
2-Ethylaniline ≥98%
2-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: –46 °C
Density: 0.983 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007751
CAS Number: 578-54-1
EINECS: 209-424-2
UN: 2273
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR