(±)-Epibromhydrin ≥98%

Danger

Synonyms: 1-Bromo-2,3-epoxypropane, (±)-3-Bromo-1,2-epoxypropane, Epibromhydrin, 3-Bromo-1,2-epoxypropane, (±)-Glycerol epibromhydrin, (±)-1-Bromo-2,3-epoxypropane

E1012-100G-A E1012-500G-A E1012-5G-A
220123-661EA 424.31 USD
220123-661 220123-662 220123-663
(±)-Epibromhydrin ≥98%
(±)-Epibromhydrin
Formula: BrH₂CCH(O)CH₂
MW: 136.98 g/mol
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: -40 °C
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
CAS Number: 3132-64-7
EINECS: 221-525-3
UN: 2558
ADR: 6.1,I

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR