EDTA tripotassium salt dihydrate

Warning

Synonyms: Ethylenedinitrilotetra-acetic acid tripotassium salt dihydrate, Tripotassium hydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid tripotassium salt dihydrate, Potassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Edetic acid tripotassium salt dihydrate

E0270-100G E0270-25G
220123-601EA 207.83 USD
220123-601 220123-602
EDTA tripotassium salt dihydrate
EDTA tripotassium salt dihydrate
Formula: C₁₀H₁₃K₃N₂O₈·2H₂O
MW: 442.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149672
CAS Number: 65501-24-8
EINECS: 241-543-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR