Diethyl malonate ≥99%

D97754-500G D97754-25G D97754-1KG D97754-2.5KG
220123-475EA 48.89 USD
220123-475 220123-474 220123-472 220123-473
Diethyl malonate ≥99%
Diethyl malonate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.055 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR