Diethyl maleate ≥97%

Warning

Synonyms: Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate

D97703-500G D97703-100G D97703-5G
220123-470EA 51.8 USD
220123-470 220123-469 220123-471
Diethyl maleate ≥97%
Diethyl maleate
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: –11 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009191
CAS Number: 141-05-9
EINECS: 205-451-9
Merck Index: 13,03150

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR