O-Dianisidine

Synonyms: 4,4'-Diamino-3,3'-dimethoxybiphenyl, 3,3'-Dimethoxybenzidine

D9143-5G D9143-25G
220123-283EA 118.92 USD
220123-283 220123-282
O-Dianisidine
O-Dianisidine
Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
MDL Number: MFCD00008372
CAS Number: 119-90-4
EINECS: 204-355-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR